Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR160 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA120396

Catalog No. PIPA120396


Add to cart

Description

Description

PA1-20396 detects G Protein-Coupled Receptor 150 in human samples. PA1-20396 has been successfully used in immunohistochemistry (paraffin) procedures. The PA1-20396 immunogen is a synthetic peptide corresponding to the 2nd extracellular domain of human G Protein-Coupled Receptor 150. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

GPR160
Polyclonal
Unconjugated
GPR160
Liquid
GPR160
Synthetic peptide corresponding to the 2nd extracellular domain of human G Protein-Coupled Receptor 150.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.01% sodium azide; pH 7.4
Q9UJ42
GPCR1, GPCR150
Rabbit
IgG
25 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
26996
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit