Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR32 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA132788

Catalog No. PIPA132788


Add to cart

Description

Description

PA1-32788 detects GPCR GPR32 from human samples. PA1-32788 has been successfully used in immunohistochemistry (paraffin tissue) applications. The PA1-32788 immunogen is: Synthetic peptide, KLH conjugated.

Specifications

Specifications

GPR32
Polyclonal
Unconjugated
GPR32
Liquid
Rabbit
IgG
25 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
2854
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.01% sodium azide; pH 7.7
O75388
GPR32
Synthetic peptide, KLH conjugated.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit