Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR35 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA120485

Catalog No. PIPA120485


Add to cart

Description

Description

PA1-20485 detects G Protein-Coupled Receptor GPR35 in Human samples. PA1-20485 has been successfully used in Immunohistochemistry (Paraffin) procedures. PA1-20485 immunogen corresponds to Synthetic peptide (aa sequence is considered to be commercially sensitive).

GPCR GPR35 is a member of the G protein coupled receptor (GPCR) family. Members of this protein family contain seven putative transmembrane domains and may mediate signaling processes to the interior of the cell via activation of heterotrimeric G proteins.
Specifications

Specifications

GPR35
Polyclonal
Unconjugated
GPR35
Liquid
Rabbit
IgG
25 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
2859
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.01% sodium azide; pH 7.7
Q9HC97
GPR35
Synthetic peptide (aa sequence is considered to be commercially sensitive)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit