Learn More
GPR4 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA112725
Catalog No. 01-674-852
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
A suggested positive control for this product is human brain lysate.
G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPR4 is a member of this family (subfamily lysophospholipid/lysosphingolipid) and its expression has been reported in kidney, heart, and especially lung, where it is at least five-fold greater than in other tissues.
Specifications
Specifications
GPR4
Polyclonal
PBS with 0.08% sodium azide
P46093
G Protein Coupled Receptor GPR4
Rabbit
IgG
10 Miniblots
-20° C, Avoid Freeze/Thaw Cycles
Primary
2828
Western Blot
Unconjugated
GPR4
Liquid
GPR4
Synthetic peptide corresponding to GPR4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human