Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR63 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA120424

Catalog No. PIPA120424

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

PA1-20424 detects G Protein-Coupled Receptor GPR63 in human samples. PA1-20424 has been successfully used in immunohistochemistry (paraffin) procedures. The PA1-20424 immunogen is a synthetic peptide corresponding to the 1st extracellular loop of human G Protein-Coupled Receptor GPR63. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

G protein-coupled receptors (GPCRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPR63 is a member of this family (subfamily: lysophospholipid/lysosphingolipid) and its expression has been reported in several brain regions, including caudate, frontal cortex, hypothalamus, midbrain, and thalamus, as well as in small intestine, stomach, and thymus.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GPR63
Polyclonal
Unconjugated
Q9BZJ6
GPR63
Synthetic peptide corresponding to the 1st extracellular loop of human G Protein-Coupled Receptor GPR63.
25 μg
Primary
Human
Antibody
IgG
Immunohistochemistry (Paraffin)
1 mg/mL
GPR63
PSP24(beta), PSP24B
Rabbit
Antigen affinity chromatography
RUO
81491
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit