Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR81 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA120524

Catalog No. PIPA120524


Add to cart

Description

Description

PA1-20524 detects G Protein-Coupled Receptor GPR81 in human samples. PA1-20524 has been successfully used in immunohistochemistry (Paraffin) procedures. The PA1-20524 immunogen is a synthetic peptide corresponding to the C-terminal portion of human G Protein-Coupled Receptor GPR81.

The G protein-coupled receptor GPCR GPR81 has been reported primarily in adipose and pituitary. ESTs have been isolated from human bladder cancer and colon cancer libraries. It has not been detected in frontal, temporal and occipital lobes of the cortex, basal forebrain, caudate nucleus, nucleus accumbens, and hippocampus.
Specifications

Specifications

GPR81
Polyclonal
Unconjugated
HCAR1
Liquid
HCAR1
Synthetic peptide corresponding to the C-terminal portion of human G Protein-Coupled Receptor GPR81.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.01% sodium azide; pH 7.7
Q9BXC0
FKSG80, GPR104, LACR1, TA-GPCR
Rabbit
IgG
25 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
27198
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit