Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR97, Polyclonal, Invitrogen™

Catalog No. PIPA132756
Change view
Click to view available options
Quantity:
25 μg
Catalog No. Quantity
PIPA132756 25 μg
1 options

Catalog No. PIPA132756

Supplier: Thermo Scientific PA132756

Rabbit Polyclonal Antibody

PA1-32756 detects GPCR GPR97 from human samples.PA1-32756 has been successfully used in immunohistochemistry (paraffin tissue) applications.The PA1-32756 immunogen is: Synthetic peptide, KLH conjugated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen G Protein-Coupled Rec. 97
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Gene ADGRG3
Gene Accession No. Q86Y34
Gene Alias PB99, PGR26
Gene Symbols ADGRG3
Host Species Rabbit
Immunogen Synthetic peptide, KLH conjugated.
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222487
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.