Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LGR6 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA133166

Catalog No. PIPA133166


Add to cart

Description

Description

PA1-33166 detects GPCR LGR6 from human samples. PA1-33166 has been successfully used in immunohistochemistry (paraffin tissue) applications. The PA1-33166 immunogen is: Synthetic peptide, KLH conjugated. Peptide sequence identity/similarity to Mouse: 65/80%.

Specifications

Specifications

LGR6
Polyclonal
Unconjugated
LGR6
Liquid
LGR6
Synthetic peptide, KLH conjugated. Peptide sequence identity/similarity to Mouse: 65/80%.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.01% sodium azide; pH 7.7
Q9HBX8
A530037C04Rik, D830026M09
Rabbit
IgG
25 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
59352
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit