Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR61, Polyclonal, Invitrogen™

Catalog No. PIPA132869
Change view
Click to view available options
Quantity:
25 μg
Catalog No. Quantity
PIPA132869 25 μg
1 options

Catalog No. PIPA132869

Supplier: Thermo Scientific PA132869

Rabbit Polyclonal Antibody

This gene belongs to the G-protein coupled receptor 1 family. G protein-coupled receptors contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. The protein encoded by this gene is most closely related to biogenic amine receptors.

PA1-32869 detects GPCR LS189885 from human samples.PA1-32869 has been successfully used in immunohistochemistry (paraffin tissue) applications.The PA1-32869 immunogen is: Synthetic peptide, KLH conjugated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen G Protein-Coupled Rec. LS189885
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Gene GPR61
Gene Accession No. Q9BZJ8
Gene Alias BALGR, GPCR3
Gene Symbols GPR61
Host Species Rabbit
Immunogen Synthetic peptide, KLH conjugated.
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83873
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.