Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR149 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA132871

Catalog No. PIPA132871


Add to cart

Description

Description

PA1-32871 detects GPCR R35/PGR10 from human samples. PA1-32871 has been successfully used in immunohistochemistry (paraffin tissue) applications. The PA1-32871 immunogen is: Synthetic peptide, KLH conjugated.

Specifications

Specifications

GPR149
Polyclonal
Unconjugated
GPR149
Liquid
GPR149
Synthetic peptide, KLH conjugated.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.01% sodium azide; pH 7.7
Q86SP6
IEDA, PGR10
Rabbit
IgG
25 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
344758
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit