Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR56 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA133156

Catalog No. PA133156


Add to cart

Description

Description

PA1-33156 detects GPCR TM7XN1/GPR56 from human samples. PA1-33156 has been successfully used in immunohistochemistry (paraffin tissue) applications. The PA1-33156 immunogen is: Synthetic peptide, KLH conjugated. Peptide sequence identity/similarity to Mouse: 75/87%. Rat: 68/87%.

Specifications

Specifications

GPR56
Polyclonal
Unconjugated
ADGRG1
Liquid
ADGRG1
Synthetic peptide, KLH conjugated. Peptide sequence identity/similarity to Mouse: 75/87%. Rat: 68/87%.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.01% sodium azide; pH 7.7
Q9Y653
Cyt28, TM7LN4, TM7XN1
Rabbit
IgG
25 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
9289
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit