Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPRC6A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521914

Catalog No. PIPA521914


Add to cart

Description

Description

PA5-21914 targets G protein-coupled receptor family C group 6 member A precursor in WB applications and shows reactivity with Human samples. The PA5-21914 immunogen is a synthetic peptide corresponding to a region within amino acids 862 and 926 of GPRC6A (Uniprot IDNo.Q5T6X5). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

GPRC6A
Polyclonal
Unconjugated
GPRC6A
Liquid
GPRC6A
Synthetic peptide corresponding to a region within amino acids 862 and 926 of GPRC6A (Uniprot IDNo.Q5T6X5)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q5T6X5
GPCR, bA86F4.3, G-protein coupled receptor GPCR33, G-protein coupled receptor family C group 6 member A, hGPCR33, hGPRC6A, predicted with SOSUI analysis, seven transmembrane helix receptor
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
222545
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit