Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gαq Mouse, Unlabeled, Clone: 10, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612705

 View more versions of this product

Catalog No. BDB612705


Add to cart

Description

Description

The GTP binding regulatory proteins (G proteins) consist of three subunits: α, β, and γ. These heterotrimeric proteins function at membranes to relay signals from cell surface receptors to intracellular effectors. The α subunit is unique for each G protein and contains the site of GTP binding and hydrolysis, as well sites for receptor and effector interactions. The βγ subunit complex interacts directly with receptors and the α subunit. The Gα family includes four families: the Gαs family including Gαs , Gαo1f, and Gαt, the Gαi family including Gαi, Gαo, and Gαz, the Gαq/Gα11 family and the Gα12/13 family. The Gαq protein is 88% homologus with Gα11 and both are widely expressed. These G proteins activate phospholipase C proteins, which induce calcium signaling events. G protein coupled receptors (GPCRs) involved in regulating Wnt signaling activate Gαq, phospholipase Cβ, and induce calcium-dependent activation of calpain. These events promote β-catenin nuclear export and proteolysis. Gαq has also been implicated in metabotropic glutamate receptor signaling. Thus, Gαq isoforms activate phospholipase C proteins in various G-protein coupled receptor pathways.

Immunofluorescence, Western Blotting

Specifications

Specifications

Gαq
10
Unconjugated
Affinity Purified
Human Gαq aa. 22-31
150μg
Cell Biology
Primary
Canine, Chicken, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.