Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LY6G6D Rabbit anti-Goat, Human, Non-human primate, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523303

Catalog No. PIPA523303


Add to cart

Description

Description

G6D, also known as Lymphocyte antigen 6 complex locus G6D(LY6G6D), is a type I transmembrane protein belonging to the immunoglobin (Ig) superfamily, which is comprised of cell-surface proteins involved in the immune system and cellular recognition. It is a disulfide linked homodimer with an Ig-like V-type domain and is known to interact with GRB2 and GRB7 in a phosphorylation-dependent manner thus playing an important role in their downstream signal transduction pathways.
Specifications

Specifications

LY6G6D
Polyclonal
Unconjugated
LY6G6D
Liquid
LY6G6D
Synthetic peptide (66 QVGRPAPDPGKPGRESR 82) of G6D  protein.
Protein A
RUO
Antibody
Polyclonal
Goat, Human, Non-human primate
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
O95868
DADB-110M10.6, C6orf23, G6D, LY6-D, MEGT1, NG25, lymphocyte antigen 6 complex locus protein G6d, lymphocyte antigen-6 G6D, megakaryocyte-enhanced gene transcript 1 protein, protein Ly6-D
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
102180269, 58530
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit