Learn More
GRB2-associated binding protein 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human GAB1 protein.
Supplier: Abnova Corporation H00002549D01P
Description
The protein encoded by this gene is a member of the IRS1-like multisubstrate docking protein family. It is an important mediator of branching tubulogenesis and plays a central role in cellular growth response, transformation and apoptosis. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: MSGGEVVCSGWLRKSPPEKKLKRYAWKRRWFVLRSGRLTGDPDVLEYYKNDHAKKPIRIIDLNLCQQVDAGLTFNKKEFENSYIFDINTIDRIFYLVADSEEEMNKWVRCICDICGFNPTEEDPVKPPGSSLQAPADLPLAINTAPPSTQADSSSATLPPPYQLINVPPHLETLGIQEDPQDYLLLINCQSKKPEPTRTHADSAKSTSSETDCNDNVPSHKNPASSQSKHGMNGFFQQQMIYDSPPSRAPSASVDSSLYNLPRSYSHDVLPKVSPSSTEADGELYVFNTPSGTSSVETQMRHVSISYDIPPTPGNTYQIPRTFPEGTLGQTSKLDTIPDIPPPRPPKPHPAHDRSPVETCSIPRTASDTDSSYCIPTAGMSPSRSNTISTVDLNKLRKDASSQDCYDIPRAFPSDRSSSLEGFHNHFKVKNVLTVGSVSSEELDENYVPMNPNSPPRQHSSSFTEPIQEANYVPMTPGTFDFSSFGMQVPPPAHMGFRSSPKTPPRRPVPVADCEPPPVDRNLKPDRKGQSPKILRLKPHGLERTDSQTIGDFATRRKVKPAPLEIKPLPEWEELQAPVRSPITRSFARDSSRFPMSPRPDSVHSTTSSSDSHDSEENYVPMNPNLSSEDPNLFGSNSLDGGSSPMIKPKGDKQVEYLDLDLDSGKSTPPRKQKSSGSGSSVADERVDYVVVDQQKTLALKSTREAWTDGRQSTESETPAKSVKSpecifications
GRB2-associated binding protein 1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human GAB1 protein. | |
GAB1 | |
GAB1 | |
GAB1 (NP_997006.1, 1 a.a. ∼ 724 a.a) full-length human protein. | |
100 μg | |
Membrane Receptors | |
2549 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
In situ PLA, Western Blot | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
NM_207123 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.