Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-GABA-A R beta 2, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP136958

Catalog No. NBP136958


Add to cart

Description

Description

GABA-A R beta 2 Polyclonal antibody specifically detects GABA-A R beta 2 in Human, Mouse, Rat, Bovine, Canine samples. It is validated for Peptide ELISA.
Specifications

Specifications

GABA-A R beta 2
0.5 mg/mL
Peptide ELISA Detection limit 1:4000
NP_068711.1
GABA(A) receptor subunit beta-2, gamma-aminobutyric acid (GABA) A receptor, beta 2, gamma-aminobutyric acid A receptor beta 2, gamma-aminobutyric acid receptor subunit beta-2, MGC119386, MGC119388, MGC119389
Goat
IgG
0.1 mg
Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary
2561
Bovine, Canine, Human, Mouse, Rat
ELISA
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
GABRB2
Peptide with sequence C-HSFGRNALERHVAQ corresponding to internal region according to NP_000804.1.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is expected to recognize both reported isoforms (NP_068711.1; NP_000804.1).
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only