Learn More
GABRG2 Rabbit anti-Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OPA104110
Catalog No. OPA104110
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This antibody is predicted to react with bovine, chicken, canine, human, mouse and non-human primate based on 100% sequence homology. In Western blot, this antibody detects a 44-46 kDa protein representing GABA-A receptor subunit gamma 2 from rat brain fractions.
Gamma-aminobutyric acid (GABA) is the primary inhibitory neurotransmitter in the central nervous system, causing a hyperpolarization of the membrane through the opening of a Cl- channel associated with the GABA-A Receptor (GABA-A R) subtype. GABA-A Rs are important therapeutic targets for a range of sedative, anxiolytic, and hypnotic agents and are implicated in several diseases including epilepsy, anxiety, depression, and substance abuse. The GABA-A R is a multimeric subunit complex. To date six alphas, four betas and four gammas, plus alternative splicing variants of some of these subunits, have been identified. Injection in oocytes or mammalian cell lines of cRNA coding for α- and β-subunits results in the expression of functional GABA A-Rs sensitive to GABA. However, coexpression of a gamma-subunit is required for benzodiazepine modulation. The various effects of the benzodiazepines in brain may also be mediated via different alpha-subunits of the receptor. Lastly, phosphorylation of beta subunits of the receptor has been shown to modulate GABA-A R function.
Specifications
Specifications
GABRG2
Polyclonal
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5
P18508
Gamma aminobutyric acid (GABA) A receptor gamma 2
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
29709
Dot Blot, Western Blot
Unconjugated
Gabrg2
Liquid
GABRG2
Fusion protein from the cytoplasmic loop of the gamma 2 subunit of rat GABRG2
Affinity chromatography
RUO
Antibody
Polyclonal
Rat