Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABRA1, Polyclonal, Invitrogen™

Catalog No. PIPA184892
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA184892 100 μL
1 options

Catalog No. PIPA184892

Supplier: Thermo Scientific PA184892

Rabbit Polyclonal Antibody

Gamma-aminobutyric acid (GABA) A receptors are GABA gated chloride channels, which are important mediators of neuronal inhibition in the mammalian central nervous system. Activation of the GABAA site results in increased intracellular chloride concentration and hyperpolarization of the cell. GABAA receptors are pentameric structures assembled from a family of distinct subunits. At least fourteen distinct subunits have been identified, which show distinct patterns of temporal and spatial expression. GABAA receptors may be modulated by several drug types including benzodiazepines and barbiturates, and by phosphorylation by protein kinase C (PKC) and protein kinase A (PKA). The GABA-A receptors are important targets for a variety of drugs such as benzodiazepines, barbiturates, steroids, anaesthetics and convulsants.

Predicted to react with bovine, chicken, dog, human and monkey based on sequence homology.In Western blot, this antibody detects a band at ∽51kDa in rat forebrain lysate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABRA1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene Gabra1
Gene Accession No. P62812, P62813
Gene Symbols GABRA1
Host Species Rabbit
Immunogen Fusion protein within the cytoplasmic loop of rat GABAA receptor alpha 1 subunit.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14394, 29705
Target Species Mouse, Rat
Content And Storage 4°C or -20°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.