Learn More
MilliporeSigma™ anti-GABA A Receptor δ-subunit, Polyclonal, (Chemicon) Non-distribution product as customer accommodation.
Manufacturer:  MilliporeSigma™ AB9752/DEL
Catalog No. 50-174-246
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Γ-aminobutyric acid (GABA) is the major inhibitory neurotransmitter in the mammalian brain where it acts at GABA-A receptors, which are ligand-gated chloride channels. Chloride conductance of these channels can be modulated by agents such as benzodiazepines that bind to the GABA-A receptor. The GABA-A receptor is generally pentameric and there are five types of subunits: α, β, γ, δ, and ρ. This gene encodes the δ subunit.
Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
Specifications
GABA A Receptor δ-subunit
Affinity Purified
Fusion protein from the N-terminal of rat GABAA Receptor δ-subunit.
RUO
Primary
Bovine, Human, Murine, Rat
Unconjugated
Rabbit
100μL
Neuroscience
Polyclonal