Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABARAPL1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526141

Catalog No. PIPA526141


Add to cart

Description

Description

Increases cell-surface expression of kappa-type opioid receptor through facilitating anterograde intracellular trafficking of the receptor.
Specifications

Specifications

GABARAPL1
Polyclonal
PBS with 0.09% sodium azide
Q9H0R8
APG8-LIKE, APG8L, ATG8, ATG8B, ATG8L, GEC1, GABA(A) receptor-associated protein-like 1, GEC-1, early estrogen-regulated protein, gamma-aminobutyric acid receptor-associated protein-like 1, glandular epithelial cell protein 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23710
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
GABARAPL1
Liquid
GABARAPL1
human GABARAPL1 recombit protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit