Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABARAPL2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527594

Catalog No. PIPA527594


Add to cart

Description

Description

PA5-27594 targets GABARAPL2 in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-27594 immunogen is recombit protein fragment corresponding to a region within amino acids 1 and 104 of Human GABARAPL2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Involved in intra-Golgi traffic. Modulates intra-Golgi transport through coupling between NSF activity and SNAREs activation. It first stimulates the ATPase activity of NSF which in turn stimulates the association with GOSR1.
Specifications

Specifications

GABARAPL2
Polyclonal
Unconjugated
GABARAPL2
Liquid
GABARAPL2
Recombit protein fragment corresponding to a region within amino acids 1 and 104 of Human GABARAPL2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 40% glycerol and 0.01% thimerosal; pH 7
P60520
ATG8, ATG8C, GATE-16, GATE16, GEF-2, GEF2, MAP1 light chain 3 related protein, MAP1 light chain 3-related protein, gamma-aminobutyric acid receptor-associated protein-like 2, ganglioside expression factor 2, general protein transport factor p16, golgi-associated ATPase enhancer of 16 kDa
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
11345
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit