Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABBR2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523720

Catalog No. PIPA523720

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with rat based on sequence homology.

The multi-pass membrane protein encoded by this gene belongs to the G-protein coupled receptor 3 family and GABA-B receptor subfamily. The GABA-B receptors inhibit neuronal activity through G protein-coupled second-messenger systems, which regulate the release of neurotransmitters, and the activity of ion channels and adenylyl cyclase. This receptor subunit forms an active heterodimeric complex with GABA-B receptor subunit 1, neither of which is effective on its own. Allelic variants of this gene have been associated with nicotine dependence.
Specifications

Specifications

GABBR2
Polyclonal
Unconjugated
GABBR2
Liquid
GABBR2
KLH conjugated synthetic peptide between 861-890 amino acids from the C-terminal region of human GABBR2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry, Western Blot
0.4 mg/mL
PBS with 0.09% sodium azide
O75899, Q80T41
GABABR2, GPR51, GPRC3B, HG20, HRIHFB2099, G protein-coupled receptor 51, G-protein coupled receptor 51, GABA-B receptor 2, GABA-B receptor, R2 subunit, GABA-B-R2, GABA-BR2, gamma-aminobutyric acid type B receptor subunit 2, gb2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
242425, 9568
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit