Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABRA4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526306

Catalog No. PIPA526306

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

GABRA4 is the major inhibitory neurotransmitter in the mammalian brain where it acts at GABA-A receptors, which are ligand-gated chloride channels. Chloride conductance of these channels can be modulated by agents such as benzodiazepines that bind to the GABA-A receptor.
Specifications

Specifications

GABRA4
Polyclonal
PBS with 0.09% sodium azide
P48169
GABA(A) receptor, alpha 4, gamma-aminobutyric acid receptor subunit alpha-4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2557
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
GABRA4
Liquid
GABRA4
KLH conjugated synthetic peptide between 350-377 amino acids from the central region of human GABRA4
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit