Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAD65 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521297

Catalog No. PIPA521297


Add to cart

Description

Description

PA5-21297 targets GAD65 in WB applications and shows reactivity with Human and mouse samples. The PA5-21297 immunogen is a synthetic peptide corresponding to a region within amino acids 465 and 558 of GAD65 (Uniprot IDNo.Q05329). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

GAD65
Polyclonal
Unconjugated
GAD2
Liquid
GAD2
Synthetic peptide corresponding to a region within amino acids 465 and 558 of GAD65 (Uniprot IDNo.Q05329)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 20% glycerol and 0.01% thimerosal; pH 7
Q05329, P48320, Q05683
RP11-420F12.2, GAD65, 65 kDa glutamic acid decarboxylase, GAD-65, Glutamate decarboxylase-2 (pancreas), glutamate decarboxylase 2, glutamate decarboxylase 65 kDa isoform
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
14417, 24380, 2572
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit