Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAD67 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141138

Catalog No. 01-675-717


Add to cart

Description

Description

The sequence used is 100% homologous with mouse and 94% homologous with rat GaD67. Suggested positive control: antigen standard for GAD1 (transient overexpression lysate), SK-N-SH.

GAD67, a 67KDa protein, is a neuronal isoform of this enzyme predomitly expressed in human Brain. It consists of Pyridoxal 5’-phosphate binding site and conserved Cysteine residues which are vital for its activity and is inhibited due to phosphorylation. It has high binding affinity for the cofactor Pyridoxal-5’phosphate and its role has been implicated in normal Palate development. It is found to be associated with childhood onset schizophrenia and craniofacial malformations and has been identified as a major auto antigen and an auto-reactive T-cell target in IDDM. Deficiency of this enzyme leads to pyridoxine dependency along with epilepsy. It also might play a role in stiff main syndrome.
Specifications

Specifications

GAD67
Polyclonal
Unconjugated
GAD1
Liquid
GAD1
Synthetic peptide, KLH-conjugated, corresponding to a portion of amino acids 1-100 of human GaD67.
Protein G
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q99259
Glutamic acid decarboxylase 67
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
2571
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit