Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GADD45G, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127707

Catalog No. PIPA127707

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Involved in the regulation of growth and apoptosis. Mediates activation of stress-responsive MTK1/MEKK4 MAPKKK. This protein is a member of a group of genes whose transcript levels are increased following stressful growth arrest conditions and treatment w

PA1-27707 detects GADD45G from human, mouse, and rat samples.PA1-27707 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The PA1-27596 immunogen is a synthetic peptide corresponding to human Growth arrest and DNA-damage-inducible protein GADD45 gamma.
Specifications

Specifications

GADD45G
Polyclonal
Unconjugated
GADD45G
Purified
GADD45G
Synthetic peptide corresponding to human Growth arrest and DNA-damage-inducible protein GADD45 gamma.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Rat
Western Blot
1mg/mL
PBS with no preservative
O95257, Q9Z111, Q9WTQ7
CR6, DDIT2, GADD45gamma, GRP17
Chicken
IgY
50μg
Store at -20°C
Primary
10912
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit