Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Gag polyprotein, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP178117

Catalog No. NBP178117


Add to cart

Description

Description

Gag polyprotein Polyclonal antibody specifically detects Gag polyprotein in Virus samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Gag polyprotein
1.22 mg/mL
Western Blot 1:1000-1:10000, ELISA 1:5000-1:20000
Affinity Purified
gag
This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids from an internal region of Gag polyprotein.
Immunogen affinity purified
RUO
Primary
This affinity purified antibody is directed against FIV Matrix Protein p15.
ELISA, Western Blot
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Pr55Gag
Rabbit
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Virus
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only