Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GALK2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513845

Catalog No. PIPA513845


Add to cart

Description

Description

GALK2 is a highly efficient N-acetylgalactosamine (GalNAc) kinase, which has galactokinase activity when galactose is present at high concentrations.
Specifications

Specifications

GALK2
Polyclonal
Unconjugated
GALK2
Liquid
GALK2
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human GALK2
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Western Blot
2 mg/mL
PBS with 0.09% sodium azide
Q01415
N-acetylgalactosamine kinase; GalNAc kinase; Galactokinase 2; GK2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2585
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit