Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GALNT2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA521541
Description
Recommended positive controls: Molt-4, mouse adipose, rat brain. Predicted reactivity: Mouse (97%), Rat (98%), Zebrafish (92%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GALNT2 Polyclonal antibody specifically detects GALNT2 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
GALNT2 | |
Polyclonal | |
Unconjugated | |
GALNT2 | |
AI480629; E4TF1B; GABP subunit beta-1; GABP subunit beta-2; GABPB; GABPB1; GABPB-1; GABPB2; GABPB-2; GalNAc-T2; galnt2; im:7140357; polypeptide GalNAc transferase 2; Polypeptide N-acetylgalactosaminyltransferase 2; Polypeptide N-acetylgalactosaminyltransferase 2 soluble form; pp-GaNTase 2; ppGaNTase-T2; protein-UDP acetylgalactosaminyltransferase 2; RP5-956O18.1; si:dkey-162b3.1; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 2; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 2; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 2 (GalNAc-T2) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
108148, 2590, 292090 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1.77 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q10471, Q6PB93 | |
GALNT2 | |
Recombinant protein fragment contain a sequence corresponding to a region within amino acids 55 and 369 (Q10471) of GALNT2 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction