Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gamma Catenin Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA137257

Catalog No. 01-673-180


Add to cart

Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is breast carcinoma.

The catenins (@alpha;, β, γ, and δ) are ubiquitously expressed, cytoplasmic proteins that associate with E-cadherin at cellular junctions. α-catenin can also bind P-cadherin and N-cadherin. γ-catenin, also known aas plakoglobin, is an 82kDa protein that binds with a-catenin and N-cadherin.
Specifications

Specifications

gamma Catenin
Polyclonal
PBS with 1% BSA and <0.1% sodium azide; pH 7.4
P14923
ARVD12, CTNNG, DP3, DPIII, PDGB, PKGB, catenin cadherin-associated protein, gamma 80kD, catenin cadherin-associated protein, gamma 80kDa, catenin gamma, desmoplakin III, desmoplakin-3, gamma-catenin
Rabbit
IgG
1 mL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
3728
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
JUP
Liquid
JUP
Synthetic peptide derived from the C-terminus of human catenin-gamma protein
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit