Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gamma Tubulin Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA138817

Catalog No. 01-674-762


Add to cart

Description

Description

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is colon carcinoma.

Tubulin gamma, an essential structural protein for cell growth, organizes microtubule arrays in the nucleus and cytoplasm. Tubulin gamma- depleted cells fail to form functional spindles and arrest during nuclear division. Tubulin gamma associates with spindle body components Spc97 and Spc98 to form the Tubulin gamma complex. The phosphrylation of Tubulin gamma at Tyr445 plays a regulatory role in microtubule formation. The incidence rate for this phosphorylation event peaks during G1.
Specifications

Specifications

gamma Tubulin
Polyclonal
PBS with 1% BSA and 0.1% sodium azide
P23258
GCP-1, TUBG, TUBGCP1
Rabbit
IgG
1 mL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
7283
Immunohistochemistry (Paraffin)
Unconjugated
TUBG1
Liquid
TUBG1
Synthetic peptide derived from C-terminus of human tubulin gamma protein
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit