Learn More
GAP43 Rabbit anti-Canine, Chicken, Human, Mouse, Non-human primate, Porcine, Rat, HRP, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA522898
Catalog No. PIPA522898
Add to cart
Description
Description
This antibody does not react with xenopus samples. Suggested positive control: embryonic nervous tissue, brain, injured nerve.
GAP43 (Neuromodulin) is associated with nerve growth. GAP43 is expressed by developing and regenerating neurons, and to a lesser extent reactive glial cells. It is a major component of the motile "growth cones" that form the tips of elongating axons. GAP43 also plays a role in axonal and dendritic filopodia induction.
Specifications
Specifications
GAP43
Polyclonal
HRP
GAP43
Liquid
GAP43
Synthetic peptide corresponding to amino acids 216-226 of rat/mouse GAP-43, (C)-KEDPEADQEHA coupled to maleimide-activated KLH.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Canine, Chicken, Human, Mouse, Non-human primate, Porcine, Rat
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
0.68 mg/mL
PBS with no preservative
P35001, P17677, P06837, P07936
B-50, PP46, axonal membrane protein GAP-43, calmodulin-binding protein P-57, growth-associated protein 43, nerve growth-related peptide GAP43, neural phosphoprotein B-50, neuromodulin, neuron growth-associated protein 43, protein F1
Rabbit
IgG
100 μL
4° C, store in dark
Primary
100623423, 14432, 2596, 29423, 427955, 478572