Learn More
GAP43 Rabbit anti-Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA136072
Catalog No. 01-672-255
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Reconstitute with 50-100 μL of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide.
GAP43 (Neuromodulin) is associated with nerve growth. GAP43 is expressed by developing and regenerating neurons, and to a lesser extent reactive glial cells. It is a major component of the motile "growth cones" that form the tips of elongating axons. GAP43 also plays a role in axonal and dendritic filopodia induction.
Specifications
Specifications
GAP43
Polyclonal
whole serum with 0.1% sodium azide
P07936
Basp2, MGC156666
Rabbit
IgG
RUO
Antibody
Polyclonal
Rat
Immunofluorescence, Immunohistochemistry (Frozen)
Unconjugated
Gap43
Lyophilized
GAP43
Synthetic GAP-43 (211-226), conjugated to BSA.
50 μL
4° C
Primary
29423