Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAS41 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA511736

Catalog No. PIPA511736

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse based on sequence homology.

NuBI-1 is found in the nucleoli. It has high sequence homology to human MLLT1, and yeast and human MLLT3 proteins. Both MLLT1 and MLLT3 proteins belong to a class of transcription factors, indicating that the encoded protein might also represent a transcription factor. This protein is thought to be required for RNA transcription. The gene for this protein has been shown to be amplified in tumors.
Specifications

Specifications

GAS41
Polyclonal
PBS with 0.09% sodium azide
O95619
YEATS domain-containing protein 4; Glioma-amplified sequence-41; Gas41; NuMA binding protein 1; NuBI-1; NuBI1; YEATS4; GAS41;
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
8089
Western Blot
Unconjugated
YEATS4
Liquid
YEATS4
KLH conjugated synthetic peptide between 16-46 amino acids from the N-terminal region of human GAS41 (YEATS4/NuBI-1)
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit