Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GASP2 Mouse anti-Human, Clone: 289802, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523992

Catalog No. PIMA523992


Add to cart

Description

Description

In direct ELISAs and Western blots, no cross-reactivity with recombit human GASP-1 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

Specifications

Specifications

GASP2
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q96NZ8
WFIKKN, WFIKKN1, C16orf12, Chromosome 16 Open Reading Frame 12, GASP2, GASP-2, Growth And Differentiation Factor-Associated Serum Protein 2, hGASP-2, RJD2, WAP Four-Disulfide Core Domain 20A, WAP, Follistatin/Kazal, Immunoglobulin, Kunitz And Netrin Domain Containing 1, WAP, FS, Ig, KU, And NTR-Containing Protein, WFDC20A, WFIKKN
Mouse
IgG2B
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
117166
Immunohistochemistry, Immunoprecipitation, Western Blot
289802
Unconjugated
WFIKKN1
Lyophilized
WFIKKN1
Mouse myeloma cell line NS0-derived recombit human GASP-2/WFIKKN Ala20-Asp548
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit