Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gastrin Guinea Pig anti-Rat, Polyclonal, Invitrogen™

Guinea Pig Polyclonal Antibody

Manufacturer:  Invitrogen PA136019

Catalog No. 01-672-209


Add to cart

Description

Description

A suggested positive control for IHC applications is rat antrum. Reconstitute with 50-100 µL of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide. Once reconstituted, store at -20°C.

Gastrin is a 17 amino acid peptide hormone and neurotransmitter widely distributed throught the gastrointestinal (GI) tract and the central nervous system (CNS). Antibodies that react specifically with gastrin may be used to study the differential tissue expression and intracellular and subcellular localization of gastrin in neuroendocrine cells of the gastrointestinal tract, and in the CNS.
Specifications

Specifications

Gastrin
Polyclonal
whole serum with 0.1% sodium azide
P04563
Gas, PPG34
Guinea Pig
IgG
RUO
Antibody
Polyclonal
Rat
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Unconjugated
Gast
Lyophilized
GAST
Synthetic human gastrin I, conjugated to BSA.
50 μL
4° C
Primary
25320
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit