Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Gastrin Rabbit anti-Human, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA516409

Catalog No. PIPA516409



Description

Description

PA5-16409 targets Gastrin in IHC (P) applications and shows reactivity with Human and Rat samples. The PA5-16409 immunogen is a Synthetic peptide from human gastrin.

Gastrin, a polypeptide hormone, occurs naturally in three forms: gastrin-14, gastrin-17 and gastrin-34.
Specifications

Specifications

Gastrin
Polyclonal
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
P01350, P04563
GAST
A Synthetic peptide from human gastrin
Purified
RUO
Antibody
Polyclonal
Human, Rat
Immunohistochemistry (Paraffin)
Unconjugated
GAST
Liquid
Rabbit
IgG
500 μL
4° C
Primary
2520, 25320
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit