Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-GATA-3, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP151912

Catalog No. NBP151912


Add to cart

Description

Description

GATA-3 Polyclonal antibody specifically detects GATA-3 in Human, Mouse samples. It is validated for Western Blot, Peptide ELISA.
Specifications

Specifications

GATA-3
0.5 mg/mL
Western Blot 0.3 - 1 ug/ml, Immunohistochemistry 3 ug/ml, Immunohistochemistry-Paraffin 3 ug/ml, Peptide ELISA Detection limit 1:64000
NP_001002295.1
131320, GATA binding protein 3, GATA-binding factor 3, GATA-binding protein 3, HDRMGC5199, MGC2346, MGC5445, trans-acting T-cell-specific transcription factor GATA-3
Goat
IgG
0.1 mg
Cancer, Innate Immunity
Primary
2625
Human, Mouse
ELISA, Western Blot
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
GATA3
Peptide with sequence EVTADQPRWVSHH-C corresponding to N-Terminus according to NP_001002295.1, NP_002042.1.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is expected to recognize both reported isoforms (NP_001002295.1 and NP_002042.1).
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only