Learn More
GATA3 Mouse, Alexa Fluor 647, Clone: L50-823, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 560078
Catalog No. BDB560078
Add to cart
Description
Description
GATA3 (GATA binding protein 3) is a member of the GATA family of transcription factors. This ∼50-kDa nuclear protein regulates the development and subsequent maintenance of multiple tissues. GATA3 is involved in the development of T lymphocytes (regulates T cell receptor subunit gene expression) and the differentiation of mature T cells to become Th2 cells. The expressed levels of normal or mutant GATA3 are also associated with the behaviors of various cancer cells including estrogen receptor-positive breast carcinoma cells.
Specifications
Specifications
GATA3
L50-823
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
IgG1, κ
RUO
Primary
Human, Murine, Rat
In vitro Assay
Alexa Fluor 647
Affinity Purified
Conserved peptide between the trans-activation and DNA-binding domains of human, mouse and rat GATA3
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal