Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATA5 Rabbit anti-Human, Polyclonal, Invitrogen™

Catalog No. PIPA1103
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIPA1103 100 μg
1 options

Catalog No. PIPA1103

Supplier: Thermo Scientific PA1103

Rabbit Polyclonal Antibody

PA1-103X has been successfully used in Western blot applications with human samples.

GATA binding factor 5 is a transcription regulator involved in embryonic development. In addition, the protein encoded by this gene contains two GATA-type zinc fingers and is known to bind to hepatocyte nuclear factor-1alpha (HNF-1alpha). This interaction is essential for cooperative activation of the intestinal lactase-phlorizin hydrolase promoter. GATA-5 may be involved in the establishment of cardiac smooth muscle cell diversity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GATA5
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide
Gene GATA5
Gene Accession No. Q9BWX5
Gene Alias GATA5, GATA binding factor 5
Gene Symbols GATA5
Host Species Rabbit
Immunogen Pooled peptides RPLVRPQKRLSSSRRA and EDDSLAPGHLEFKFEPED
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140628
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.