Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-GBA Clone: 200,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ AP1140100UG
Description
Specifically detects GBA Clone: 200 in Human samples, and it is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immuno Blotting
Acid beta-glucosidase (GBA) is a lysosomal membrane protein that cleaves the beta-glucosidic linkage of glycosylceramide, an intermediate in glycolipid metabolism. It catalyzes the breakdown of the glycolipid glucosylceramide to ceramide and glucose. Mutations in this gene cause Gaucher disease, a lysosomal storage disease characterized by an accumulation of glucocerebrosides.Specifications
GBA | |
Monoclonal | |
Unconjugated | |
Mouse | |
100 μg | |
Primary | |
Human | |
Purified |
ELISA, Immunoblot, Immunoblot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
200 | |
In PBS, pH 7.2. | |
A recombinant polypeptide corresponding to amino acids of 146-236 human GBA, expressed as a GST fusion protein | |
RUO | |
Recognizes the ∽60kDa GBA protein in MCF-7 and HeLa cells and human breast cancer tissue. | |
−20°C | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction