Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GBAS Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer: Invitrogen PA525911
Description
Chromosomal region 7p12, which contains GBAS, is amplified in approximately 40% of glioblastomas, the most common and maligt form of central nervous system tumor.The predicted 286-amino acid protein contains a signal peptide, a transmembrane domain, and 2 tyrosine phosphorylation sites. The GBAS transcript is expressed most abundantly in heart and skeletal muscle. GBAS protein might be involved in vesicular transport.Specifications
GBAS | |
Polyclonal | |
Liquid | |
NIPSNAP2 | |
NIPSNAP2, 4-nitrophenylphosphatase domain and non-neuronal SNAP25-like 2, protein NipSnap homolog 2 | |
NIPSNAP2 | |
KLH conjugated synthetic peptide between 130-159 amino acids from the central region of human GBAS | |
Polyclonal | |
Antigen affinity chromatography | |
RUO | |
Human |
Flow Cytometry,Immunohistochemistry,Western Blot | |
Unconjugated | |
PBS with 0.09% sodium azide | |
O75323 | |
2631 | |
Rabbit | |
IgG | |
Primary | |
400 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Antibody |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title