Learn More
GNB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSG00020W
Catalog No. PIOSG00020W
Add to cart
Description
Description
Glycerol (1:1) may be added for additional stability.
Heterotrimeric guanine nucleotide-binding proteins (G proteins), which integrate signals between receptors and effector proteins, are composed of an alpha, a beta, and a gamma subunit. These subunits are encoded by families of related genes. This gene encodes a beta subunit. Beta subunits are important regulators of alpha subunits, as well as of certain signal transduction receptors and effectors. This gene uses alternative polyadenylation signals.
Specifications
Specifications
GNB1
Polyclonal
whole serum with no preservative
P62873, P62874, P54311
Transducin beta chain 1, GNB1, Guanine nucleotide binding protein subunit beta-1
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry, Western Blot
Unconjugated
GNB1
Lyophilized
GNB1
A synthetic peptide from the internal region of human GBB1
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
14688, 24400, 2782