Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GCAP3 Monoclonal Antibody (3F10), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA520796
Description
Peptide Sequence: KNKLLDMFMA VQALNGQQTL SPEEFINLVF HKIDINNDGE LTLEEFINGM AKDQDLLEIV YKSFDFSNVL RVICNGKQPD METDSSKSPD KAGLGKVKMK
GCAP3 Monoclonal antibody specifically detects GCAP3 in Human samples. It is validated for ELISA, Immunoprecipitation, Western Blot.Specifications
GCAP3 | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
O95843 | |
GUCA1C | |
GUCA1C (NP_005450.2, 110 a.a. ∽ 209 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Immunoprecipitation, Western Blot | |
3F10 | |
Unconjugated | |
GUCA1C | |
GCAP 3; GCAP3; Guanylate cyclase activator 1C; guanylyl cyclase-activating protein 3; GUCA1C; Hu-DF1-VAR | |
Mouse | |
Affinity chromatography | |
RUO | |
9626 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction