Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCF2 Mouse anti-Human, Unlabeled, Clone: 32, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612161

 View more versions of this product

Catalog No. BDB612161


Add to cart

Description

Description

Regulation of transcription is mediated by transcription factors that bind regulatory elements in gene promotors and enhancers, and alter the rate of transcription. Transcriptional repressors reduce transcription by their intrinsic repressor domain activity, or by binding to sites in the regulatory domain where a co-repressor can bind. GC-binding factors (GCF and GCF2) are transcriptional repressors that bind promoters with GC-rich sequences. GCF represses transcription of EGF, TGFα, and insulin-like growth factor II recepetors. GCF2 has some homology with GCF, and has also been identified as TRIP and LRRFIP1. The sequence of GCF2 includes an RNA binding domain (RBD), and N-glycosylation site, and a nuclear localization signal (NLS). GCF2 is widely expressed in many tissues and cells. GCF2 binds the EGFR and PDGFR promoters, and represses both EGFR and PDGFR transcription. In addition, GCF2 interacts with the leucine-rich repeat of flightless I, a protein that binds actin and contains a gelsolin-like domain. Thus, GCF2 may have functions in both RNA-cytoskeleton interactions, as well as roles in transcriptional repression.

Immunofluorescence, Western Blotting

Specifications

Specifications

GCF2
32
Unconjugated
Affinity Purified
Human GCF2 aa. 210-415
150μg
Cell Biology
Primary
Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.