Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCH1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525735

Catalog No. PIPA525735


Add to cart

Description

Description

This gene encodes a member of the GTP cyclohydrolase family. The encoded protein is the first and rate-limiting enzyme in tetrahydrobiopterin biosynthesis, catalyzing the conversion of GTP into 7,8-dihydroneopterin triphosphate. BH4 is an essential cofactor required by aromatic amino acid hydroxylases as well as nitric oxide synthases. Mutations in this gene are associated with maligt hyperphenylalaninemia and dopa-responsive dystonia. Several alternatively spliced transcript variants encoding different isoforms have been described; however, not all variants give rise to a functional enzyme.
Specifications

Specifications

GCH1
Polyclonal
PBS with 0.09% sodium azide
P30793
DYT14, DYT5, DYT5a, GCH, GTP-CH-1, GTPCH1, HPABH4B, GTP cyclohydrolase I, GTP-CH-I, dystonia 14, guanosine 5′-triphosphate cyclohydrolase I
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2643
Immunohistochemistry, Western Blot
Unconjugated
GCH1
Liquid
GCH1
KLH conjugated synthetic peptide between 39-67 amino acids from the N-terminal region of human GCH1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit