Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYF2, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA131763

Catalog No. PIPA131763

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a nuclear protein that interacts with cyclin D-type binding-protein 1, which is thought to be a cell cycle regulator at the G1/S transition. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.

PA1-31763 detects GCIP interacting p29 from human samples.PA1-31763 has been successfully used in Western blot applications.The PA1-31763 immunogen is: Peptide with sequence AEIKQNLERGTAV, from C Terminus of the protein sequence according to NP_056299.
Specifications

Specifications

GCIP interacting p29
Polyclonal
Unconjugated
SYF2
Purified
SYF2
Peptide with sequence AEIKQNLERGTAV, from C Terminus of the protein sequence according to NP_056299.
IgG fraction
RUO
Antibody
Polyclonal
Human, Murine, Rat
Western Blot
0.5mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q9D198, Q4QRB2, O95926
CBPIN, DKFZp564O2082, NTC31, P29, fSAP29
Goat
IgG
50μg
Avoid Freeze/Thaw Cycles
Primary
25949
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit