Learn More
GCLM Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA526111
Catalog No. PIPA526111
Add to cart
Description
Description
Gamma-glutamylcysteine synthetase (gamma-GCS) is the rate limiting enzyme for glutathione (L-gamma-glutamyl-L-cysteinylglycine, GSH) synthesis. GSH is ubiquitous in mammalian cells as a vital intra- and extracellular protective antioxidant. Gamma-GCS is a heterodimer of a heavy catalytic subunit and a light regulatory subunit that is responsive to inflammation, phenolic ntioxidants, heat shock, oxidants and cytokines.
Specifications
Specifications
GCLM
Polyclonal
Unconjugated
GCLM
Liquid
GCLM
KLH conjugated synthetic peptide between 246-274 amino acids from the C-terminal region of human GCLM
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.4 mg/mL
PBS with 0.09% sodium azide
P48507, O09172
GLCLR, GCS light chain, GSC light chain, gamma-ECS regulatory subunit, gamma-glutamylcysteine synthetase regulatory subunit, glutamate--cysteine ligase modifier subunit, glutamate--cysteine ligase regulatory subunit, glutamate-cysteine ligase (gamma-glutamylcysteine s
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
14630, 2730