Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCN5 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527146

Catalog No. PIPA527146


Add to cart

Description

Description

PA5-27146 targets GCN5 in WB applications and shows reactivity with Human samples. The PA5-27146 immunogen is synthetic peptide corresponding to a region within amino acids 775 and 837 of Human GCN5. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

KAT2A, or GCN5, is a histone acetyltransferase (HAT) that functions primarily as a transcriptional activator. It also functions as a repressor of NF-kappa-B (see MIM 164011) by promoting ubiquitination of the NF-kappa-B subunit RELA (MIM 164014) in a HAT-independent manner (Mao et al., 2009 [PubMed 19339690]).
Specifications

Specifications

GCN5
Polyclonal
Unconjugated
KAT2A
Liquid
KAT2A
Synthetic peptide corresponding to a region within amino acids 775 and 837 of Human GCN5
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Q92830
GCN5, GCN5L2, PCAF-b, hGCN5, GCN5 (general control of amino-acid synthesis, yeast, homolog)-like 2, General control of amino acid synthesis, yeast, homolog-like 2, STAF97, general control of amino acid synthesis protein 5-like 2, histone acetyltransferase GCN5, histone acetyltransferase K
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
2648
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit