Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCP2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA521433

Catalog No. PIPA521433


Add to Cart

Description

Description

Recommended positive controls: A431, HeLa, NIH-3T3. Predicted reactivity: Mouse (88%), Rat (88%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

GCP2 Polyclonal antibody specifically detects GCP2 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GCP2
Polyclonal
Unconjugated
TUBGCP2
1700022B05Rik; a disintegrin and metalloprotease domain 8; Adam8; Gamma-ring complex protein 103 kDa; gamma-tubulin complex component 2; GCP2; GCP-2; Grip103; h103p; hGCP2; hGrip103; hSpc97; SPBC97; Spc97p; spindle pole body protein Spc97 homolog; Tubgcp2; tubulin gamma complex associated protein 2; tubulin, gamma complex associated protein 2
Rabbit
Antigen affinity chromatography
RUO
10844, 74237
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q921G8, Q9BSJ2
TUBGCP2
Recombinant fragment corresponding to a region within amino acids 1 and 226 of GCP2 (Uniprot ID#Q9BSJ2)
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit